Orders LinkContactUsLinkComponent
J.T.Baker®, Hematology Controls, 3-diff Control Extended
3-Different control extended, medium, 2,5 mL
Catalog # 3422.
Supplier:   Avantor
J.T.Baker®, Hematology Controls, 3-diff Control Extended
Catalog # 3422.
Supplier:  Avantor
CAS Number:  
J.T.Baker®, Hematology Controls, 3-diff Control Extended
3-Different control extended, medium, 2,5 mL
CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 2,5 mL
Product Details & Documents
In vitro diagnostic hematology control product, intended for professional use only.
 : Parameter Controls are to be stored upright at 2-8°C when not in use. Stored at this temperature, the Controls are guaranteed stable until the expiry date.
Specifications
Specifications
  • Pk:
    2,5 mL
  • 説明:
    3-Different control extended, medium
Specifications