Orders LinkContactUsLinkComponent
VWR®, 遠心分離機用チューブ
15 ml, Screw cap, 3000×g, sterile, 50/bag, 10 bags/case
Catalog # 525-0640
   Low Manufacturing Impact
Supplier:   VWR Collection
VWR®, 遠心分離機用チューブ
Catalog # 525-0640
Supplier:  VWR Collection
   Low Manufacturing Impact
VWR®, 遠心分離機用チューブ
VWR®, 遠心分離機用チューブ
15 ml, Screw cap, 3000×g, sterile, 50/bag, 10 bags/case
Price
Inquire For Price
Log in for availability
1 * 500 items
Product Details & Documents
PS, clear, conical, with flat cap with integral sealing.
  • Electron beam irradiated
  • Longer length screw caps with sealing ring prevent any leakage
  • Engraved graduations at the conical bottom of each tube
  • Graduations and writing area are chloroform-resistant
  • During the freezing process, it is better to use plastic rack
  • During high-speed centrifugation, it is better to use suitable rotor adapters
Specifications
  • Closure type:
    Screw cap
  • Packed:
    50/bag, 10 bags/case
  • Version:
    sterile
  • 容量:
    15 ml
  • Seq:
    03
  • RCF max.:
    3000×g
  • Cat. No.:
    525-0640
  • Environmental certification:
    ISO 14001/Certificate No.: 04923E00243R0M
  • Environmentally Preferable:
    Low Manufacturing Impact