Orders LinkContactUsLinkComponent
スライドホルダ、VWR®製
スライドホルダ、VWR®製
Double slide holders, Clear, PP, 2
  VWRU95042-668
スライドホルダ、VWR®製
  VWRU95042-668
スライドホルダ、VWR®製
スライドホルダ、VWR®製
Double slide holders, Clear, PP, 2
$104.36
1 * 250 items
Polypropylene resistant slide holders easy to open for durable protection.
Certifications: FDA certified
Packaging: 250 slides per box, four boxes per case.
  • No. of slides:
    2
  • Colour:
    Clear
  • 説明:
    Double slide holders
  • Material:
    PP
  • Seq:
    02