ビーカー, 使い捨て, PP
15 ml, 5-15 ml
Catalog # 414004-143
Supplier: VWR Collection
Jump to:
Product Details & Documents
PP, transparent.
- Withstand temperatures up to 121 °C
- Shatter-resistant
- Easy-to-read graduations
- Transparent for clear content visibility
Beakers feature a pouring spout for quick and easy dispensing. Highly resistant to strong acids. Stackable. Steam autoclavable.
注意: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Specifications
- 目盛り範囲:5-15 ml
- 容量:15 ml
- Seq:01