静電気防止用ブラシ、 StaticAway™ (スタティックアウェイ
Long handle brush, Natural bristle, Plastic, 50,8 mm
Catalog # VWRU89094-718
Supplier: VWR Collection
Jump to:
Product Details & Documents
Brushes clean by dissipating static and whisking away harmful dust, lint, and dirt. Ideal for cleaning balance pans, optics, cuvettes, slides, film, microscopes, electronics, and computer monitors. The compact brush is suitable for electronic balances, providing accurate results by eliminating pan weight variances caused by static.
- Natural bristles will not scratch critical surfaces
- Brushes drain static and clean with a unique blend of natural hair, synthetic conductive fiber, and a conductive composite handle
- Long handle brushes allow access to difficult places
Specifications
- Bristle material:Natural bristle
- Handle material:Plastic
- 説明:Long handle brush
- Seq:02
- 総長:50,8 mm