Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR® Basic, Bouffant Caps, Latex-Free

VWR® Basic, Bouffant Caps, Latex-Free

Blue, 610 mm
Catalog # VWRU89131-622
Supplier:   VWR Collection
VWR® Basic, Bouffant Caps, Latex-Free
Catalog # VWRU89131-622
Supplier:  VWR Collection
VWR® Basic, Bouffant Caps, Latex-Free
VWR® Basic, Bouffant Caps, Latex-Free

Blue, 610 mm

Price
Inquire For Price
Log in for availability
1 * 1.000 items
Product Details & Documents

These disposable caps are made from a non woven, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.

  • Free from natural rubber latex
  • Available in a wide range of colours

Weight: 10 g/m²

包装: 100 pieces per bag, 10 bags per case.

Specifications
  • 直径:
    610 mm
  • Colour:
    Blue
  • Seq:
    05