Orders LinkContactUsLinkComponent
作業着、ジッパー付き 最大
作業着、ジッパー付き 最大
White, 4XL
Catalog # VWRU414004-701
Supplier:   VWR Collection
作業着、ジッパー付き 最大
Catalog # VWRU414004-701
Supplier:  VWR Collection
作業着、ジッパー付き 最大
作業着、ジッパー付き 最大
White, 4XL
Price
Inquire For Price
Log in for availability
1 * 25 items
Product Details & Documents
VWR® Maximum SF frocks are the cleanest and most durable within the entire VWR® clothing line. Made from spunbonded polypropylene fabric (SPP) and coated with breathable polyethylene film, these garments have been designed to optimise barrier protection, cleanliness and comfort properties.
  • Snap collar, raglan sleeves, zip closure, elasticated cuffs
  • Superior fluid and particulate barrier
  • Lowest level of particle shedding: Level 1 Helmke drum classification
  • Light chemical splash resistance
  • Improved breathability to ensure user comfort
  • Soft, cloth-like fabric with maximum tear strength
The frocks provide barrier protection against fluids, biologicals and particulates. A high vapour transmission rate keeps the wearer comfortable, even throughout extended use. Appropriate for industrial, pharmaceutical, food processing, construction, and certain environmental clean-up applications.
Certifications: Tested and manufactured in an ISO certified facility under stringent process controls. Lot-controlled and validated through independent laboratory testing.
Specifications
  • 衣服のサイズ:
    4XL
  • Colour:
    White
  • Gender:
    Unisex
  • Closure type:
    Zip closure
  • Garment application:
    Cleanroom|Laboratory
  • Cat. No.:
    VWRU414004-701
  • Pockets:
    Without pockets