作業着、ジッパー付き 最大
Product Details & Documents
VWR® Maximum SF frocks are the cleanest and most durable within the entire VWR® clothing line. Made from spunbonded polypropylene fabric (SPP) and coated with breathable polyethylene film, these garments have been designed to optimise barrier protection, cleanliness and comfort properties.
- Snap collar, raglan sleeves, zip closure, elasticated cuffs
- Superior fluid and particulate barrier
- Lowest level of particle shedding: Level 1 Helmke drum classification
- Light chemical splash resistance
- Improved breathability to ensure user comfort
- Soft, cloth-like fabric with maximum tear strength
The frocks provide barrier protection against fluids, biologicals and particulates. A high vapour transmission rate keeps the wearer comfortable, even throughout extended use. Appropriate for industrial, pharmaceutical, food processing, construction, and certain environmental clean-up applications.
Certifications: Tested and manufactured in an ISO certified facility under stringent process controls. Lot-controlled and validated through independent laboratory testing.
Specifications
- 衣服のサイズ:4XL
- Colour:White
- Gender:Unisex
- Closure type:Zip closure
- Garment application:Cleanroom|Laboratory
- Cat. No.:VWRU414004-701
- Pockets:Without pockets