ブーツ付きオーバーオール, SMS
White, L
Catalog # VWRU414004-689
Supplier: VWR Collection
Jump to:
Product Details & Documents
These overalls with integrated shoe covers are manufactured from a unique multilayered spunbonded-meltblown-spunbonded (SMS) fabric that offers high tensile strength while remaining soft, comfortable and breathable. The material is low-linting and resistant to tears and punctures.
- Light fluid and particulate barrier
- Excellent breathability
- Inset sleeves, front zipper, tapered collar
- Elastic cuffs, ankles, and back
- No pockets
- Serged seams
These static-dissipative garments act as a fluid and particulate barrier and provide light chemical splash resistance. Ideal for use in high-class protection areas of medical, industrial, laboratory, electronic, and pharmaceutical environments.
Certifications: Tested and manufactured in an ISO certified facility under stringent process controls. Lot-controlled and validated through independent laboratory testing.
Specifications
- 衣服のサイズ:L
- Colour:White
- Seq:03
- Cat. No.:VWRU414004-689
- Garment application:Laboratory
- Bacterial filtration efficiency:0,9