Orders LinkContactUsLinkComponent
ブーツ付きオーバーオール, SMS
White, M
Catalog # VWRU414004-688
Supplier:   VWR Collection
ブーツ付きオーバーオール, SMS
Catalog # VWRU414004-688
Supplier:  VWR Collection
ブーツ付きオーバーオール, SMS
White, M
Price
Inquire For Price
Log in for availability
1 * 25 items
Product Details & Documents
These overalls with integrated shoe covers are manufactured from a unique multilayered spunbonded-meltblown-spunbonded (SMS) fabric that offers high tensile strength while remaining soft, comfortable and breathable. The material is low-linting and resistant to tears and punctures.
  • Light fluid and particulate barrier
  • Excellent breathability
  • Inset sleeves, front zipper, tapered collar
  • Elastic cuffs, ankles, and back
  • No pockets
  • Serged seams
These static-dissipative garments act as a fluid and particulate barrier and provide light chemical splash resistance. Ideal for use in high-class protection areas of medical, industrial, laboratory, electronic, and pharmaceutical environments.
 : Tested and manufactured in an ISO certified facility under stringent process controls. Lot-controlled and validated through independent laboratory testing.
Specifications
  • 衣服のサイズ:
    M
  • Colour:
    White
  • Seq:
    02
  • Cat. No.:
    VWRU414004-688
  • Garment application:
    Laboratory
  • Bacterial filtration efficiency:
    0,9