保護ガウン、SPP
Yellow, XL
VWRU414004-468
Economical gowns are manufactured from a strong, lightweight, breathable and non-linting spunbonded polypropylene (SPP) fabric.
- Provide first line barrier
- Elastic cuffs and ties at the neck and waist ensure secure closure with
- Serged seams
- No pockets
Gowns protect against dirt, grime and certain dry particulates in nonhazardous environments.
: Tested and manufactured in an ISO certified facility under stringent process controls. Lot-controlled and validated through independent laboratory testing.
- 衣服のサイズ:XL
- Colour:Yellow
- Seq:02
- Seam type:Serged
- Cat. No.:VWRU414004-468
- Garment application:Medical|Clinical
- Pockets:Without pockets
- Fabric:SPP