Orders LinkContactUsLinkComponent
保護ガウン、SPP
Yellow, XL
  VWRU414004-468
保護ガウン、SPP
  VWRU414004-468
保護ガウン、SPP
保護ガウン、SPP
Yellow, XL
1 * 50 items
Economical gowns are manufactured from a strong, lightweight, breathable and non-linting spunbonded polypropylene (SPP) fabric.
  • Provide first line barrier
  • Elastic cuffs and ties at the neck and waist ensure secure closure with
  • Serged seams
  • No pockets
Gowns protect against dirt, grime and certain dry particulates in nonhazardous environments.
 : Tested and manufactured in an ISO certified facility under stringent process controls. Lot-controlled and validated through independent laboratory testing.
  • 衣服のサイズ:
    XL
  • Colour:
    Yellow
  • Seq:
    02
  • Seam type:
    Serged
  • Cat. No.:
    VWRU414004-468
  • Garment application:
    Medical|Clinical
  • Pockets:
    Without pockets
  • Fabric:
    SPP