Orders LinkContactUsLinkComponent
保護ガウン、SPP
保護ガウン、SPP
Yellow, L
Catalog # VWRU414004-467
Supplier:   VWR Collection
保護ガウン、SPP
Catalog # VWRU414004-467
Supplier:  VWR Collection
保護ガウン、SPP
保護ガウン、SPP
Yellow, L
Price
Inquire For Price
Log in for availability
1 * 50 items
Product Details & Documents
Economical gowns are manufactured from a strong, lightweight, breathable and non-linting spunbonded polypropylene (SPP) fabric.
  • Provide first line barrier
  • Elastic cuffs and ties at the neck and waist ensure secure closure with
  • Serged seams
  • No pockets
Gowns protect against dirt, grime and certain dry particulates in nonhazardous environments.
Certifications: Tested and manufactured in an ISO certified facility under stringent process controls. Lot-controlled and validated through independent laboratory testing.
Specifications
  • 衣服のサイズ:
    L
  • Colour:
    Yellow
  • Seq:
    01
  • Seam type:
    Serged
  • Cat. No.:
    VWRU414004-467
  • Garment application:
    Medical|Clinical
  • Pockets:
    Without pockets
  • Fabric:
    SPP