カラー標準液 ASTM
ASTM Colour Standard Sample A1, Glass bottle, 100 mL
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
Ready to use colour solutions that are prepared gravimetrically on a weight/weight basis.
- Use either as calibration and/or quality control standards
- Designed specifically for use in ASTM analytical methods, and produced in accordance with ASTM D1500, D6045, D1209
- Traceable product certified by an independent laboratory
- Supplied in high quality, tamper-evident bottles
Specifications
Specifications
- パックタイプ:Glass bottle
- Pk:100 mL
- 説明:ASTM Colour Standard Sample A1