Orders LinkContactUsLinkComponent
ビーカー, 使い捨て, PP
ビーカー, 使い捨て, PP
50 ml, 5-50 ml
Catalog # 414004-145
Supplier:   VWR Collection
ビーカー, 使い捨て, PP
Catalog # 414004-145
Supplier:  VWR Collection
ビーカー, 使い捨て, PP
ビーカー, 使い捨て, PP
50 ml, 5-50 ml
Price
$46.13
Log in for availability
1 * 100 items
Product Details & Documents
PP, transparent.
  • Withstand temperatures up to 121 °C
  • Shatter-resistant
  • Easy-to-read graduations
  • Transparent for clear content visibility
Beakers feature a pouring spout for quick and easy dispensing. Highly resistant to strong acids. Stackable. Steam autoclavable.
注意: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Specifications
  • 目盛り範囲:
    5-50 ml
  • 容量:
    50 ml
  • Seq:
    03