VWR®, 鉗子
Non sterile, 120 mm, 30 bags of 50
Supplier: VWR Collection
Jump to:
Product Details & Documents
PS, translucent blue.
- Combines high mechanical strength, flexibility and chemical resistance
- Available in sterile (individually wrapped) and non sterile versions
Documents
CoC VWR Forceps, PS, translucent blue plastic with pointed end.pdf CoC VWR Forceps, PS, Translucent Blue Plastic with Pointed Ends.pdf CoC VWR Forceps.pdf CoC VWR, Forceps.pdf CoC_VWR_FORCEPS_BLUE_TRANSLUCENT.pdf
Conformity declaration
COC_VWRI232-0191_20180910 CoC_VWRI232-0191_20190320 COC_VWRI232-0191_20190610 CoC_VWRI232-0191_20190820 COC_VWRI232-0191_20190920
More
TDS_VWR_FORCEPS_BLUE_TRANSLUCENT.pdf
Product specification
VWRI232-0191_TDS_EN
Datasheet
VWR® Lab bric-a-brac
Product Literature
Specifications
- Packed:30 bags of 50
- Type:Non sterile
- 長さ:120 mm
- Seq:02
- Cat. No.:232-0192
- Environmental certification:ISO 14001 (Certificate Registration No.131120)
- Environmentally Preferable:Low Manufacturing Impact