Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR®, 研削ジャー

VWR®, 研削ジャー

25 ml, Stainless steel, 6 mm, 10 ml
  412-0171
VWR®, 研削ジャー
  412-0171
VWR®, 研削ジャー
VWR®, 研削ジャー

25 ml, Stainless steel, 6 mm, 10 ml

 
 
   
1 * 2 items
 
 
  • 容量:
    25 ml
  • Material:
    Stainless steel
  • Seq:
    04
  • Max. feed size:
    6 mm
  • Sample volume:
    10 ml
Complementary Products

 

  •  
    説明
     
     
  • 412-0167
    Beater
       
    1 * 1 items - $0.00