Order Entry
Japan
Orders LinkContactUsLinkComponent
 

VWR®, 研削ジャー

1,5 ml, Stainless steel, 1 mm, 0,5 ml
  412-0168
VWR®, 研削ジャー
  412-0168
VWR®, 研削ジャー
VWR®, 研削ジャー

1,5 ml, Stainless steel, 1 mm, 0,5 ml

 
 
   
1 * 2 items
 
  • 容量:
    1,5 ml
  • Material:
    Stainless steel
  • Seq:
    01
  • Max. feed size:
    1 mm
  • Sample volume:
    0,5 ml
Complementary Products

 

  •  
    説明
     
     
  • 412-0167
    Beater
       
    1 * 1 items - $0.00