VWR® Q Path®, スライドトレイ
Slide tray with lid, Blue, Plastic, 20
Catalog # 631-1078
Supplier: VWR Collection
Jump to:
Product Details & Documents
Flip-top trays designed for the safe storage of up to 20 microscope slides. With or without lid.
Colour: White or blue
Documents
VWR® for Microscopy
Product Literature
Specifications
- No. of slides:20
- Colour:Blue
- 説明:Slide tray with lid
- Material:Plastic
- Seq:02