Orders LinkContactUsLinkComponent
VWR® Q Path®, スライドトレイ
VWR® Q Path®, スライドトレイ
Slide trays, without lid, White, Plastic, 20
Catalog # 631-1077
Supplier:   VWR Collection
VWR® Q Path®, スライドトレイ
Catalog # 631-1077
Supplier:  VWR Collection
VWR® Q Path®, スライドトレイ
VWR® Q Path®, スライドトレイ
Slide trays, without lid, White, Plastic, 20
Price
Inquire For Price
Log in for availability
1 * 10 items
Product Details & Documents
Flip-top trays designed for the safe storage of up to 20 microscope slides. With or without lid.
Colour: White or blue
Specifications
  • No. of slides:
    20
  • Colour:
    White
  • 説明:
    Slide trays, without lid
  • Material:
    Plastic
  • Seq:
    03