VWR® Q Path®, スライドホルダー
Slide holders, Translucent, PP, 3
631-1075
Slide holders designed for applications in clinical cytology.
- Fast, easy clip closure
- For two or three slides, depending on the model
- Modern, user friendly design
- No. of slides:3
- Colour:Translucent
- 説明:Slide holders
- Material:PP
- Seq:002