VWR® Q Path®, スライドオーガナイザー
Metal organiser with metal slide drawers (14), Green, 480×480×140 mm, 5500
631-1069
These organisers enable the archiving of a large number of microscope slides.
- Robust and stackable
W×D×H: 480×480×140 mm
: Supplied with locking rack.
- No. of slides:5500
- Colour:Green
- 説明:Metal organiser with metal slide drawers (14)
- Seq:02
- W×D×H:480×480×140 mm