Orders LinkContactUsLinkComponent
カラー標準液 ASTM
カラー標準液 ASTM
ASTM Colour Standard Sample A5, Glass bottle, 500 mL
Catalog # 84797.260
Supplier:   VWR Chemicals
カラー標準液 ASTM
Catalog # 84797.260
Supplier:  VWR Chemicals
CAS Number:  
カラー標準液 ASTM
カラー標準液 ASTM
ASTM Colour Standard Sample A5, Glass bottle, 500 mL
CAS Number:  
Price
$790.02
Log in for availability
1 * 500 mL
Product Details & Documents
Ready to use colour solutions that are prepared gravimetrically on a weight/weight basis.
  • Use either as calibration and/or quality control standards
  • Designed specifically for use in ASTM analytical methods, and produced in accordance with ASTM D1500, D6045, D1209
  • Traceable product certified by an independent laboratory
  • Supplied in high quality, tamper-evident bottles
Documents
Certificates
Search by Lot Number
Specifications
Specifications
  • パックタイプ:
    Glass bottle
  • Pk:
    500 mL
  • 説明:
    ASTM Colour Standard Sample A5
Specifications