VWR® VisiScope®, Camera Adapters
C-mount adapter compatible with Nikon microscopes, 0,5×
Catalog # 630-1856
Supplier: VWR Collection
Jump to:
Product Details & Documents
Documents
Specifications
- 説明:C-mount adapter compatible with Nikon microscopes, 0,5×
- Seq:06