シューズカバー、PP/PE、アドバンスド
Blue, 159 mm, 394 mm
Catalog # 414004-649
Supplier: VWR Collection
Jump to:
Product Details & Documents
These shoe covers are manufactured from a specially formulated PP/PE microporous film. They provide excellent fluid and barrier protection.
- Highly durable, long-lasting abrasion- and skid-resistance
- Excellent particulate barrier
- High coefficient of friction provides excellent traction
- Non linting
- Serged seams
Specifications
- Colour:Blue
- 高さ:159 mm
- 長さ:394 mm
- Cat. No.:414004-649
- Garment application:Laboratory