シューズカバー、PP/PE、アドバンスド
Blue, 171 mm, 432 mm
Catalog # 414004-650
Supplier: VWR Collection
Jump to:
Product Details & Documents
These shoe covers are manufactured from a specially formulated PP/PE microporous film. They provide excellent fluid and barrier protection.
- Highly durable, long-lasting abrasion- and skid-resistance
- Excellent particulate barrier
- High coefficient of friction provides excellent traction
- Non linting
- Serged seams
Specifications
- Colour:Blue
- 高さ:171 mm
- 長さ:432 mm
- Cat. No.:414004-650
- Garment application:Laboratory