オーバーオール、VWRシグネチャー
Overalls with elastic ankles, White, 3XL
Catalog # VWRU89012-832
Supplier: VWR Collection
Jump to:
Product Details & Documents
These protective, durable, and comfortable overalls made with DuPont™ Tyvek® IsoClean® material provide an excellent barrier against dry particulates, non hazardous liquids, and microorganisms. Garments are chemically and biologically inert, low-linting, and static-free until laundered.
- Excellent dry particulate, nonhazardous liquid, and microorganism barrier
- Chemically and biologically inert
- Low-linting, static-free
- Serged seams, laydown collar, front zipper closure, and elastic wrists
- Available with elastic ancles or attached Gripper™ boots
- Boot attachments combine an embossed PVC exterior sole with a coarsely textured interior to ensure a secure, non slip fit and feel
Specifications
- 衣服のサイズ:3XL
- Colour:White
- 説明:Overalls with elastic ankles
- Seq:06
- Cat. No.:VWRU89012-832
- Anti-static:+
- Garment application:Laboratory|Industrial