Order Entry
Japan
Orders LinkContactUsLinkComponent
 

VWR® Basic, Bouffant Caps, Latex-Free

Blue, 530 mm
Catalog # VWRU89131-620
Supplier:   VWR Collection
VWR® Basic, Bouffant Caps, Latex-Free
Catalog # VWRU89131-620
Supplier:  VWR Collection
VWR® Basic, Bouffant Caps, Latex-Free
VWR® Basic, Bouffant Caps, Latex-Free

Blue, 530 mm

Price
Inquire For Price
Log in for availability
1 * 1.000 items
Product Details & Documents

These disposable caps are made from a non woven, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.

  • Free from natural rubber latex
  • Available in a wide range of colours

Weight: 10 g/m²

Packaging: 100 pieces per bag, 10 bags per case.

Specifications
  • 直径:
    530 mm
  • Colour:
    Blue
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.