Orders LinkContactUsLinkComponent
VWR®, 超高精度K型プローブ
Air/gas type K probe model TK 401, for deep frozen goods with plastic handle (135 mm), cable (1 m) and DIN-SMP connection, −65…+600 °C, 6×130 mm
  620-1942
VWR®, 超高精度K型プローブ
  620-1942
VWR®, 超高精度K型プローブ
Air/gas type K probe model TK 401, for deep frozen goods with plastic handle (135 mm), cable (1 m) and DIN-SMP connection, −65…+600 °C, 6×130 mm
1 * 1 items
  • Range:
    −65…+600 °C
  • 説明:
    Air/gas type K probe model TK 401, for deep frozen goods with plastic handle (135 mm), cable (1 m) and DIN-SMP connection
  • 直径×長さ:
    6×130 mm