VWR®, 超高精度K型プローブ
Air/gas type K probe model TK 401, for deep frozen goods with plastic handle (135 mm), cable (1 m) and DIN-SMP connection, −65…+600 °C, 6×130 mm
620-1942
- Range:−65…+600 °C
- 説明:Air/gas type K probe model TK 401, for deep frozen goods with plastic handle (135 mm), cable (1 m) and DIN-SMP connection
- 直径×長さ:6×130 mm