VWR®, 超高精度K型プローブ
Penetration probe type K model TK 101 SC, for deep frozen goods with plastic handle (135 mm), cable (1 m), DIN-SMP connection, −65…+200 °C, 3×130 mm
Catalog # 620-1943
Supplier: VWR Collection
Jump to:
Product Details & Documents
Documents
VWR® for Temperature Measurement and Recording
Product Literature
Specifications
- Range:−65…+200 °C
- 説明:Penetration probe type K model TK 101 SC, for deep frozen goods with plastic handle (135 mm), cable (1 m), DIN-SMP connection
- 直径×長さ:3×130 mm