Orders LinkContactUsLinkComponent
VWR®, 超高精度K型プローブ
VWR®, 超高精度K型プローブ
Penetration probe type K model TK 101 SC, for deep frozen goods with plastic handle (135 mm), cable (1 m), DIN-SMP connection, −65…+200 °C, 3×130 mm
Catalog # 620-1943
Supplier:   VWR Collection
VWR®, 超高精度K型プローブ
Catalog # 620-1943
Supplier:  VWR Collection
VWR®, 超高精度K型プローブ
Penetration probe type K model TK 101 SC, for deep frozen goods with plastic handle (135 mm), cable (1 m), DIN-SMP connection, −65…+200 °C, 3×130 mm
Specifications
  • Range:
    −65…+200 °C
  • 説明:
    Penetration probe type K model TK 101 SC, for deep frozen goods with plastic handle (135 mm), cable (1 m), DIN-SMP connection
  • 直径×長さ:
    3×130 mm