Orders LinkContactUsLinkComponent
VWR®, 超高精度K型プローブ
Penetration probe type K model TK 101, with plastic handle (135 mm), cable (1 m) and DIN-SMP connection, −65…+1000 °C, 3×130 mm
  620-1940
VWR®, 超高精度K型プローブ
  620-1940
VWR®, 超高精度K型プローブ
Penetration probe type K model TK 101, with plastic handle (135 mm), cable (1 m) and DIN-SMP connection, −65…+1000 °C, 3×130 mm
1 * 1 items
  • Range:
    −65…+1000 °C
  • 説明:
    Penetration probe type K model TK 101, with plastic handle (135 mm), cable (1 m) and DIN-SMP connection
  • 直径×長さ:
    3×130 mm