Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR®, 超高精度K型プローブ

VWR®, 超高精度K型プローブ

Penetration probe type K for soft media TK 100 with DIN-SMP connection, −65…+400 °C, 1,5×100 mm
Catalog # 620-1936
Supplier:   VWR Collection
VWR®, 超高精度K型プローブ
Catalog # 620-1936
Supplier:  VWR Collection
VWR®, 超高精度K型プローブ

Penetration probe type K for soft media TK 100 with DIN-SMP connection, −65…+400 °C, 1,5×100 mm

Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Range:
    −65…+400 °C
  • 説明:
    Penetration probe type K for soft media TK 100 with DIN-SMP connection
  • 直径×長さ:
    1,5×100 mm
  • Cat. No.:
    620-1936
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.