Orders LinkContactUsLinkComponent
VWR®, 超高精度K型プローブ
General purpose probe type K, for surface, immersion, air and gas model TK 501 with plastic handle, silicone cable (1 m) and DIN-SMP connection, −65…+400 °C, 1,5×100 mm
  620-1937
VWR®, 超高精度K型プローブ
  620-1937
VWR®, 超高精度K型プローブ
General purpose probe type K, for surface, immersion, air and gas model TK 501 with plastic handle, silicone cable (1 m) and DIN-SMP connection, −65…+400 °C, 1,5×100 mm
1 * 1 items
  • Range:
    −65…+400 °C
  • 説明:
    General purpose probe type K, for surface, immersion, air and gas model TK 501 with plastic handle, silicone cable (1 m) and DIN-SMP connection
  • Seq:
    04
  • 直径×長さ:
    1,5×100 mm