VWR®, 超高精度K型プローブ
Universal flexible type K probe, model TK 430, for quick response in air, liquids, very small surfaces, twisted pair of PTFE-insulated thermowell wires and DIN-SMP connection, −65…+300 °C, 1×1000 mm
Catalog # 620-1941
Supplier: VWR Collection
Jump to:
Product Details & Documents
Documents
VWR® for Temperature Measurement and Recording
Product Literature
Specifications
- Range:−65…+300 °C
- 説明:Universal flexible type K probe, model TK 430, for quick response in air, liquids, very small surfaces, twisted pair of PTFE-insulated thermowell wires and DIN-SMP connection
- 直径×長さ:1×1000 mm