Orders LinkContactUsLinkComponent
VWR®, 超高精度K型プローブ
VWR®, 超高精度K型プローブ
Universal flexible type K probe, model TK 430, for quick response in air, liquids, very small surfaces, twisted pair of PTFE-insulated thermowell wires and DIN-SMP connection, −65…+300 °C, 1×1000 mm
Catalog # 620-1941
Supplier:   VWR Collection
VWR®, 超高精度K型プローブ
Catalog # 620-1941
Supplier:  VWR Collection
VWR®, 超高精度K型プローブ
Universal flexible type K probe, model TK 430, for quick response in air, liquids, very small surfaces, twisted pair of PTFE-insulated thermowell wires and DIN-SMP connection, −65…+300 °C, 1×1000 mm
Specifications
  • Range:
    −65…+300 °C
  • 説明:
    Universal flexible type K probe, model TK 430, for quick response in air, liquids, very small surfaces, twisted pair of PTFE-insulated thermowell wires and DIN-SMP connection
  • 直径×長さ:
    1×1000 mm