VWR®, セルスクレーパーおよびセルリフター
Cell lifter, blade width 20 mm, 250 mm
Catalog # 734-2603
Supplier: VWR Collection
Jump to:
Product Details & Documents
Specifically designed to make collecting cells easier and more effective. Available with a choice of blade positions: scraper for use in flasks, or lifter for use in harvesting cells (especially stem cells) in dishes.
- Cross-ribbed handle in ABS provides greater rigidity and ensures better control while scraping cells
- Thin, flexible TPE blade prevents damage to cells
- Individually wrapped
- Sterilised by electron beam irradiation
- Free from DNase, RNase and non-pyrogenic
Documents
Avantor Cell Culture Solutions.pdf
Product Literature
CoC_VWRI734-2602_170416-233 CoC_VWRI734-2602_180408233 CoC_VWRI734-2602_181006233 CoC_VWRI734-2602_190416233 CoC_VWRI734-2602_200424233
More
TDS VWR Cell Scraper
Datasheet
TDS_VWR_CELLSCRAPERS
Product specification
Specifications
- 説明:Cell lifter, blade width 20 mm
- Seq:002
- 総長:250 mm
- Cat. No.:734-2603
- Environmental certification:ISO 14001/Certificate No.: 04923E00243R0M
- Environmentally Preferable:Low Manufacturing Impact