VWR天秤用密度キット
Product Details & Documents
Complete kits that, together with the appropriate balance, allow the user to calculate the density of liquids (such as beverages) or non absorbent solids (rocks, polymers, gems, glass, metals etc). The density wizard function is already included as a built-in function of the balance (except for SE models: 611-2605, 611-2606, 611-2607, 611-3236, 611-3246, 611-3299, 611-3344). Density kits are not designed for use with moisture analysers. Both kits contain similar items, the difference is the balance cone attachment:
- Measuring glass cup 150 mm high, Ø 50 mm, 250 ml
- Measuring glass cup 95 mm high, Ø 65 mm, 250 ml
- Thermometer
- Glass sinker
- Sample holder for floating/non floating samples
- Hanging arm for sample holder or sinker
- Sample holder/sinker support
- Measuring glass cup support
Density kit DT002 for VWR® SM, SMG, LA, LAG, LP, LPG, LPW, LPWG, LPC, LPCG, SE or TS, TA, TP, TPW or TPC series of analytical balances with readability of 0,01; 0,1 or 1 mg.
Density kit DT004 for VWR® LP, LPG, SE or TP and TP series precision balances with readability of 0,01 g.
Documents
Manual VWR Precision Balance with Graphic Display Manual_VWR_Balances_611-3279_and_-3281 Manual_VWR_Balances_Top_load_611-2605_to_-2607,_-3236,_-3246,_-3299 Manual_VWR_Density_kit_for_solids_and_liquids_GRAF_611-2550_to_-2553 Manual_VWR_Density_kit_for_solids_and_liquids_LCD_611-2550_to_-2553
Manual
VWR® for Weighing
Product Literature
Specifications
- 説明:Density kit DT002 (solids and liquids) for VWR balances with readability 0,001; 0,0001 or 0,00001 g (1; 0,1 or 0,01 mg)
- Seq:02