Orders LinkContactUsLinkComponent
VWR®, スタンダードキュベット
Cell, 30 mm self masking continuous flowthrough quartz cell, beam height 15 mm, 1 pcs
Catalog # 634-0697
Supplier:   VWR Collection
VWR®, スタンダードキュベット
Catalog # 634-0697
Supplier:  VWR Collection
VWR®, スタンダードキュベット
Cell, 30 mm self masking continuous flowthrough quartz cell, beam height 15 mm, 1 pcs
Specifications
  • 説明:
    Cell, 30 mm self masking continuous flowthrough quartz cell, beam height 15 mm, 1 pcs
  • Seq:
    8