Order Entry
Japan
Orders LinkContactUsLinkComponent
 

ピペット用バルブ, VWR® (英語)

Dropper bulbs, 1 ml, Black
Catalog # VWRU15001-362
Supplier:   VWR Collection
ピペット用バルブ, VWR® (英語)
Catalog # VWRU15001-362
Supplier:  VWR Collection
ピペット用バルブ, VWR® (英語)
ピペット用バルブ, VWR® (英語)

Dropper bulbs, 1 ml, Black

Price
Inquire For Price
Log in for availability
1 * 12 items
Product Details & Documents

Pipette bulbs made of natural rubber.

  • Durable material
  • Bulbs are suitable for medicine droppers and small dropping pipettes
  • Neck is grooved to take flat flange of pipettes
  • Heavy-walled, cylindrical in shape
  • Bulbs fit pipettes 6 to 8 mm Ext.Ø
Specifications
  • Colour:
    Black
  • 説明:
    Dropper bulbs, 1 ml
  • Seq:
    01