Büchner製ファンネル
1860 ml, 185 mm
Catalog # VWRU89038-132
Supplier: VWR Collection
Jump to:
Product Details & Documents
Porcelain, glased inside and out, except rim.
- Resistant to chemicals
- Very good temperature resistance
- For paper or membrane filtration discs
Specifications
- 容量:1860 ml
- Seq:07
- For filter Ø:185 mm