Orders LinkContactUsLinkComponent
VWR®, シューズカバー、CPE
VWR®, シューズカバー、CPE
White, U*
Catalog # VWRU414004-513
Supplier:   VWR Collection
VWR®, シューズカバー、CPE
Catalog # VWRU414004-513
Supplier:  VWR Collection
VWR®, シューズカバー、CPE
VWR®, シューズカバー、CPE
White, U*
Price
Inquire For Price
Log in for availability
1 * 1.000 items
Product Details & Documents
Basic shoe covers manufactured from a low-density chlorinated polyethylene fabric, which is designed to protect against dirt, grime, and liquid splash.
  • CPE is water resistant and provides good resistance against chemicals
  • Economical and lint-free
Ideal for laboratory use.
* One size fits all
Specifications
  • 靴のサイズ:
    U*
  • Colour:
    White
  • Seq:
    02
  • Cat. No.:
    VWRU414004-513
  • Garment application:
    Laboratory