VWR®, 分離漏斗、Squibb
100 ml, 19/26 NS
Supplier: VWR Collection
Jump to:
Product Details & Documents
Borosilicate 3.3 glass with NS-PE stopper.
- With or without graduations
- Stopcock with PTFE key
- Very good chemical and temperature resistance
Specifications
- Socket size:19/26 NS
- 容量:100 ml
- Seq:08