VWR®, ポリエステルシールフィルム
Polyester sealing film, Transparent, 49,5 µm, -
Catalog # 731-0319
Supplier: VWR Collection
Jump to:
Product Details & Documents
Polyester film, acrylic adhesive. These 49,5 µm thick polyester films minimise evaporation and prevent spillage and contamination between wells.
- Recommended temperature range from −40 to +120 °C
- Available sterile and non sterile
- Marginally pierceable by single-channel pipette
- Applications: Short-term storage, incubation, ELISA
Each film L×D: 141,0×79,4 mm
Each film L×D: 141,0×79,4 mm
Specifications
- 光学特性:Transparent
- Sterile:-
- 説明:Polyester sealing film
- 厚さ:49,5 µm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



