Orders LinkContactUsLinkComponent
VWR®, 漏斗、粉体
150 mm, 136,5 mm, 26,7 mm, 33,0 mm
  VWRU414004-272
VWR®, 漏斗、粉体
  VWRU414004-272
VWR®, 漏斗、粉体
VWR®, 漏斗、粉体
150 mm, 136,5 mm, 26,7 mm, 33,0 mm
$41.49
1 * 4 items
PP, translucent.
  • Parallel stem minimises powder bridging
  • External ribs for better air evacuation
  • Excellent chemical and temperature resistance
  • Autoclavable
Used for transferring powders. Also suitable for liquids.
  • Funnel O-Ø:
    150 mm
  • 長さ:
    136,5 mm
  • Seq:
    06
  • Stem O-Ø:
    26,7 mm
  • Stem length:
    33,0 mm