VWR®, バルブピペット、1マーク、クラスAS
3 ml, 0,010 ± ml, 350 mm
Borosilicate 9.1 glass.
- Calibrated to deliver (TD)
- Ring mark and inscriptions in high contrast amber stain
- Not available with individual certificate
Certifications: DIN EN ISO 648
Ordering information: Dated batch identification and batch certificate included.
- 容量:3 ml
- 長さ:350 mm
- Seq:03
- Tolerance:0,010 ± ml