VWR®, バルブピペット、1マーク、クラスAS
25 ml, 0,030 ± ml, 530 mm
Supplier: VWR Collection
Jump to:
Product Details & Documents
Borosilicate 9.1 glass.
- Calibrated to deliver (TD)
- Ring mark and inscriptions in high contrast amber stain
- Not available with individual certificate
: DIN EN ISO 648
: Dated batch identification and batch certificate included.
Specifications
- 容量:25 ml
- 長さ:530 mm
- Seq:08
- Tolerance:0,030 ± ml