Orders LinkContactUsLinkComponent
VWR®, バルブピペット、1マーク、クラスAS
VWR®, バルブピペット、1マーク、クラスAS
2 ml, 0,010 ± ml, 330 mm
Catalog # 612-4130
Supplier:   VWR Collection
VWR®, バルブピペット、1マーク、クラスAS
Catalog # 612-4130
Supplier:  VWR Collection
VWR®, バルブピペット、1マーク、クラスAS
VWR®, バルブピペット、1マーク、クラスAS
2 ml, 0,010 ± ml, 330 mm
Price
Inquire For Price
Log in for availability
1 * 3 items
Product Details & Documents
Borosilicate 9.1 glass.
  • Calibrated to deliver (TD)
  • Ring mark and inscriptions in high contrast amber stain
  • Not available with individual certificate
 : DIN EN ISO 648
 : Dated batch identification and batch certificate included.
Specifications
  • 容量:
    2 ml
  • 長さ:
    330 mm
  • Seq:
    02
  • Tolerance:
    0,010 ± ml