VWR®, オーバーオール、フードまたはカラー付き
White, XL
113-1985
These disposable overalls made of PP are suitable for use in non hazardous environments.
- With hood or collar
- Zip closure
- With finger loops
Weight: 40 g/m²
- 衣服のサイズ:XL
- Colour:White
- Seq:03
- Cat. No.:113-1985
- Garment application:General Purpose|Laboratory|Industrial
- Weight:40 g/m²