VWR®, オーバーオール、フードまたはカラー付き
White, 3XL
Catalog # 113-1982
Supplier: VWR Collection
Jump to:
Product Details & Documents
These disposable overalls made of PP are suitable for use in non hazardous environments.
- With hood or collar
- Zip closure
- With finger loops
Weight: 40 g/m²
Specifications
- 衣服のサイズ:3XL
- Colour:White
- Seq:09
- Cat. No.:113-1982
- Garment application:General Purpose|Laboratory|Industrial
- Weight:40 g/m²