VWR®, バイオハザード標本袋
Biohazard specimen bags, LDPE, 305×229 mm, —
Catalog # VWRU11215-682
Supplier: VWR Collection
Jump to:
Product Details & Documents
Designed for safe and secure transport of a specimen container and documentation, these 51 µm thick bags feature a pouch to keep the paperwork separate from the container. They have a self-sealing closure and are tamper evident.
Documents
Specifications
- 説明:Biohazard specimen bags
- 長さ×幅:305×229 mm
- Material:LDPE
- Seq:04
- 厚さ:—
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



