VWR®, 乳鉢、乳棒なし
70 ml, 42 mm, 81 mm
410-0109
Porcelain, produced according to DIN 12906.
- With spout
- Glazed outside, grinding surface unglazed
- Highly resistant to acids and alkalis
- 内径:81 mm
- 容量:70 ml
- 高さ:42 mm
Porcelain, produced according to DIN 12906.