VWR®, 乳鉢、乳棒なし
1000 ml, 104 mm, 185 mm
410-0116
Porcelain, produced according to DIN 12906.
- With spout
- Glazed outside, grinding surface unglazed
- Highly resistant to acids and alkalis
- 内径:185 mm
- 容量:1000 ml
- 高さ:104 mm
Porcelain, produced according to DIN 12906.