VWR®, 乳鉢、乳棒なし
4300 ml, 160 mm, 265 mm
Catalog # 410-0118
Supplier: VWR Collection
Jump to:
Product Details & Documents
Porcelain, produced according to DIN 12906.
- With spout
- Glazed outside, grinding surface unglazed
- Highly resistant to acids and alkalis
Specifications
- 内径:265 mm
- 容量:4300 ml
- 高さ:160 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



